missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cinosulfuron, MilliporeSigma™ Supelco™

Catalog No. 111015949
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015949 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015949 Supplier Merck Emd Millipore Supplier No. 37893100MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 94593-91-6
Flash Point >144°C - Closed Cup
Physical Form Neat
Linear Formula C15H19N5O7 S
MDL Number MFCD01311813
Melting Point 140°C to 145°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 413.41
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH