missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Chlorpyrifos-(diethyl-d10), ≥97.5% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 285138-81-0 |
| Product Line | PESTANAL |
| Linear Formula | C9D10 HCl3 NO3 PS |
| Synonym | O,O-Di(ethyl-d5) O-(3,5,6-trichloropyridin-2-yl) thiophosphate; Chlorpyriphos-d10 |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 360.65 |
| Percent Purity | ≥97.5% (GC) |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH