missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Chlornitrofen, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1836-77-7 |
| Product Line | PESTANAL |
| Purity Grade Notes | C-NMR and H-NMR conform to structure |
| Linear Formula | C12H6 Cl3N1O3 |
| MDL Number | MFCD00210255 |
| Synonym | 1,3,5-Trichloro-2-(4-nitrophenoxy)benzene; 4-Nitrophenyl 2,4,6-trichlorophenyl ether |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 318.54 |
| Percent Purity | ≥98% (HPLC) |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH