missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Chlorfluazuron, MilliporeSigma™ Supelco™

Catalog No. 111015330
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015330 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015330 Supplier Merck Emd Millipore Supplier No. 3653050MGR
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 71422-67-8
Physical Form Neat
Linear Formula C20H9 Cl3F5N3O3
MDL Number MFCD00143948
Type Analytical Standard
Quantity 50 mg
Formula Weight 540.65
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH