missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Chlorfenvinphos, MilliporeSigma™ Supelco™

Catalog No. 111019467
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019467 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019467 Supplier Merck Emd Millipore Supplier No. 36551250MG

Chlorfenvinphos is an organophosphorus insecticide used for the control of the bee parasite Varroa destructor and also found to be effective against a variety of insects affecting vegetables, fruits and crops.

Agriculture

Specifications

Product Line PESTANAL
CAS 470-90-6
Physical Form Neat
Linear Formula C12H14 Cl3O4 P
MDL Number MFCD00041818
Type Analytical Standard
Quantity 250 mg
Formula Weight 359.57
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH