missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ γ-Chlordane, MilliporeSigma™ Supelco™

Catalog No. 111019926
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019926 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019926 Supplier Merck Emd Millipore Supplier No. 442599

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

γ-Chlordane belongs to the class of organochlorine pesticides (OCPs) typically utilized for agriculture applications.

Agriculture

Specifications

Packaging Ampule
Product Line PESTANAL
CAS 5103-74-2
Physical Form Neat
Type Analytical Standard
Quantity 50 mg
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH