missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Cartap hydrochloride, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 15263-52-2 |
| Physical Form | Neat |
| Linear Formula | C7H15N3O2S2 · HCl |
| MDL Number | MFCD01311812 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 273.80 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH