missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Carfentrazone-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111019023
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019023 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019023 Supplier Merck Emd Millipore Supplier No. 3407910MG
Only null left

Carfentrazone-ethyl is an aryl triazolinone reduced-risk herbicide used in the cultivation of wheat, soybean and corn.

Agriculture

Specifications

Product Line PESTANAL
CAS 128639-02-1
Physical Form Neat
Linear Formula C15H14 Cl2F3N3O3
MDL Number MFCD03792738
Synonym Ethyl 2-chloro-3-{2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1 H-1,2,4-triazol-1-yl]-4-fluoro-phenyl}propionate; Ethyl-alpha,2-dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1 H-1,2,4-triazol-1-yl]-4-fluorobenzenepropanoate
Type Analytical Standard
Quantity 10 mg
Formula Weight 412.19
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH