missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Carbofuran-d3, MilliporeSigma™ Supelco™

Catalog No. 111018245
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018245 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018245 Supplier Merck Emd Millipore Supplier No. 3401910MGR
Only null left

Carbofuran-d3 is a deuterated analog of the pesticide, carbofuran.

Agriculture

Specifications

Product Line PESTANAL
CAS 1007459-98-4
Physical Form Neat
Linear Formula C12H12D3 NO3
MDL Number MFCD07366666
Synonym 2,3-Dihydro-2,2-dimethyl-7-benzofuranol N-methylcarbamate-d3
Type Analytical Standard
Quantity 10 mg
Formula Weight 224.27
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH