missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Carbofuran-3-hydroxy, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 16655-82-6 |
| Physical Form | Neat |
| Linear Formula | C12H15 NO4 |
| MDL Number | MFCD00143168 |
| Melting Point | 140°C to 148°C |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 237.25 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH