missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Captafol, MilliporeSigma™ Supelco™

Catalog No. 111020505
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020505 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020505 Supplier Merck Emd Millipore Supplier No. 45365250MG
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 2425-06-1
Flash Point >100°C
Physical Form Neat
Linear Formula C10H9 Cl4 NO2 S
MDL Number MFCD00041816
Type Analytical Standard
Quantity 250 mg
Formula Weight 349.06
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH