missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Butylat, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 2008-41-5 |
| Physical Form | Neat |
| Linear Formula | C11H23 NOS |
| MDL Number | MFCD00055338 |
| Synonym | S-Ethyl N,N-diisobutylthiocarbamate |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 217.37 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH