missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Butachlor ESA sodium salt, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 1173022-75-7 |
| Physical Form | Neat |
| Linear Formula | C17H26 NNaO5 S |
| MDL Number | MFCD09842835 |
| Synonym | 2-[(2,6-Diethylphenyl)(butoxymethyl)amino]-2-oxo-ethanesulfonic acid sodium salt |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 379.45 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH