missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Buprofezin, MilliporeSigma™ Supelco™

Catalog No. 111015255
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015255 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015255 Supplier Merck Emd Millipore Supplier No. 37886100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buprofezin is an insect growth regulator insecticide. Its mode of action involves inhibition of chitin biosynthesis and is also known to inhibit molting against coleopteran, hemipteran insects and acarina.

Agriculture

Specifications

Product Line PESTANAL
CAS 69327-76-0
Physical Form Neat
Linear Formula C16H23N3 OS
MDL Number MFCD01681899
Type Analytical Standard
Quantity 100 mg
Formula Weight 305.44
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH