missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Bromacil, MilliporeSigma™ Supelco™

Catalog No. 111020463
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020463 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020463 Supplier Merck Emd Millipore Supplier No. 45350250MG

Bromacil is an active ingredient of the commercially marketed pesticide, Krovar, widely used for weed control in citrus orchards.

Agriculture

Specifications

Product Line PESTANAL
CAS 314-40-9
Physical Form Neat
Linear Formula C9H13 BrN2O2
MDL Number MFCD00055364
Synonym 5-Bromo-3-sec.-butyl-6-methyluracil
Type Analytical Standard
Quantity 250 mg
Formula Weight 261.12
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH