missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Brodifacoum, MilliporeSigma™ Supelco™

Catalog No. 111019370
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019370 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019370 Supplier Merck Emd Millipore Supplier No. 46036100MG

Brodifacoum is an anticoagulant rodenticide, widely used for the control of rodent population via inhibition of the vitamin K epoxide reductase, causing blood clotting alterations, leading to haemorrhaging as the ultimate cause of death.

Agriculture

Specifications

Product Line PESTANAL
CAS 56073-10-0
Physical Form Neat
Linear Formula C31H23 BrO3
MDL Number MFCD00084803
Type Analytical Standard
Quantity 100 mg
Formula Weight 523.42
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH