missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Boscalid, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 188425-85-6 |
| Physical Form | Solid |
| Linear Formula | C18H12 Cl2N2 O |
| MDL Number | MFCD06795150 |
| Synonym | 2-Chloro-N-(4 ′-chlorobiphenyl-2- yl)-nicotinamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 343.21 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH