missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Bistrifluron, Analytical Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Fused Cone |
| CAS | 201593-84-2 |
| Product Line | PESTANAL |
| Physical Form | Neat |
| Linear Formula | C16H7ClF8N2O2 |
| MDL Number | MFCD22581210 |
| Synonym | 1-[2-Chloro-3,5-bis(trifluoromethyl)phenyl]-3-(2,6-difluorobenzoyl)urea |
| Quantity | 25 mg |
| Formula Weight | 446.68 |
| Percent Purity | ≥98.0% (HPLC) |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH