missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Bispyribac sodium salt, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 125401-92-5 |
| Physical Form | Neat |
| Linear Formula | C19H17N4 NaO8 |
| MDL Number | MFCD02314071 |
| Synonym | Sodium 2,6-bis(4,6-dimethoxy-2-pyrimidinyloxy)benzoate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 452.35 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH