missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Biphenyl, MilliporeSigma™ Supelco™

Catalog No. 111020669
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020669 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020669 Supplier Merck Emd Millipore Supplier No. 358001G
Agriculture and Environmental

Specifications

Product Line PESTANAL
Ignition Point 1004°F
CAS 92-52-4
Vapor Pressure 9.46 mmHg (115°C)
Vapor Density 5.31 (vs. air)
Flash Point 110°C
Physical Form Neat
Linear Formula C6H5C6H5
MDL Number MFCD00003054
Melting Point 68°C to 70°C
Boiling Point 255°C
Type Analytical Standard
Quantity 1 g
Formula Weight 154.21
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH