missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ γ-BHC (Lindane) 0.1 μg/mL in Hexane, Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | PESTANAL |
| Concentration | 0.1 μg/mL in hexane |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | PESTANAL™ Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH