missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Benomyl, MilliporeSigma™ Supelco™

Catalog No. 111018877
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018877 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018877 Supplier Merck Emd Millipore Supplier No. 45339250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benomyl is characterized under the benzimidazole fungicide and is known to suppress fungal growth in a wide variety of plants. Its mode of action involves inhibition of tubulin polymerization.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 17804-35-2
Physical Form Neat
Linear Formula C14H18N4O3
MDL Number MFCD00084636
Melting Point >300°C
Synonym Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate; Benomyl; NSC 263489
Type Analytical Standard
Quantity 250 mg
Formula Weight 290.32
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH