missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Azoxystrobin, MilliporeSigma™ Supelco™

Catalog No. 111019138
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019138 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019138 Supplier Merck Emd Millipore Supplier No. 31697100MG
Only null left

Azoxystrobin belongs to strobilurin class of fungicides which is based on lead molecule strobilurin A. Its target site is mitochondrial respiration of fungi.

Agriculture

Specifications

Product Line PESTANAL
CAS 131860-33-8
Physical Form Neat
Linear Formula C22H17N3O5
MDL Number MFCD08277047
Type Analytical Standard
Quantity 100 mg
Formula Weight 403.39
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH