missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Azaconazole, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 60207-31-0 |
| Physical Form | Neat |
| Linear Formula | C12H11 Cl2N3O2 |
| MDL Number | MFCD00865600 |
| Synonym | 1-[2-(2,4-Dichlorophenyl)-1,3-dioxolan-2-ylmethyl]-1 H-1,2,4-triazole |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 300.14 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH