missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Anilofos, MilliporeSigma™ Supelco™

Catalog No. 111019468
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019468 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019468 Supplier Merck Emd Millipore Supplier No. 37876100MG

Anilofos is a pre-emergence, organophosphorus herbicide, which is commonly used for the control of various annual grass weeds and sedges such as barnyard grasses in rice-based cropping system. Its mode of action involves the inhibition of cell division stage in weeds.

Agriculture

Specifications

Product Line PESTANAL
CAS 64249-01-0
Flash Point >100°C
Physical Form Neat
Linear Formula C13H19 ClNO3 PS2
MDL Number MFCD01311803
Synonym S-[2-(4-Chloro-N-isopropylanilino)-2-oxoethyl]-O,O′-dimethyl dithiophosphate
Type Analytical Standard
Quantity 100 mg
Formula Weight 367.85
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH