missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Amitrol, MilliporeSigma™ Supelco™

Numéro de catalogue. 111020003
Modifier l'affichage
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111020003 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111020003 Fournisseur Merck Emd Millipore Code fournisseur 45324250MG
Il en reste null
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Spécifications

Product Line PESTANAL
CAS 61-82-5
Physical Form Neat
Linear Formula C2H4N4
MDL Number MFCD00005230
Melting Point 150°C to 153°C
Synonym 3-Amino-1,2,4-triazole; 1,2,4-Triazol-3-amine; 3-AT; Amitrol
Type Analytical Standard
Quantity 250 mg
Formula Weight 84.08
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH