missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Aminopyralid, MilliporeSigma™ Supelco™

Catalog No. 111013194
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013194 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013194 Supplier Merck Emd Millipore Supplier No. 3245725MG

It has been used to control Canada Thistle (Cirsium arvense), a weed in a variety of crops.

Agriculture

Specifications

Product Line PESTANAL
CAS 150114-71-9
Physical Form Neat
Linear Formula C6H4 Cl2N2O2
MDL Number MFCD08273857
Synonym 4-Amino-3,6-dichloropicolinic acid; 4-Amino-3,6-dichloropyridine-2-carboxylic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 207.01
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH