missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Aminoethoxyvinyl glycine hydrochloride, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 55720-26-8 |
| Physical Form | Neat |
| Linear Formula | C6H12N2O3 · HCl |
| MDL Number | MFCD00043020 |
| Synonym | (S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride; L-alpha-(2-Aminoethoxyvinyl)glycine hydrochloride; AVG-HCl |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 196.63 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH