missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Alloxydim-sodium, MilliporeSigma™ Supelco™

Catalog No. 111014606
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014606 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014606 Supplier Merck Emd Millipore Supplier No. 45319250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture

Specifications

Product Line PESTANAL
CAS 55635-13-7
Flash Point >100°C
Physical Form Neat
Linear Formula C17H24 NNaO5
MDL Number MFCD16657007
Type Analytical Standard
Quantity 250 mg
Formula Weight 345.37
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH