missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Allethrin, MilliporeSigma™ Supelco™

Catalog No. 111019839
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019839 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019839 Supplier Merck Emd Millipore Supplier No. 33396100MG

Allethrin is a non-cyano containing pyrethroid household insecticide, that can be used to control flies and mosquitoes.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 584-79-2
Physical Form Neat
Linear Formula C19H26O3
MDL Number MFCD00045443
Synonym Allylcinerine
Type Analytical Standard
Quantity 100 mg
Formula Weight 302.41
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH