missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Aldrin Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 309-00-2 |
| Linear Formula | C12H8 Cl6 |
| MDL Number | MFCD00135587 |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 364.91 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL in acetonitrile |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH