missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Aldicarb-(N-methyl-13C,d3 carbamoyl-13C), ≥98.5% (HPLC), MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 1261170-75-5 |
| Flash Point | >100°C |
| Linear Formula | 13C2C5D3H11N2O2 S |
| MDL Number | MFCD17677362 |
| Synonym | (1 E)-2-Methyl-2-(methylthio)-O-[(methyl-13 C-d3-amino)carbonyl-13 C]oxime propanal |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 195.27 |
| Percent Purity | ≥98.5% (HPLC) |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH