missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Alanycarb, MilliporeSigma™ Supelco™

Catalog No. 111018291
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018291 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018291 Supplier Merck Emd Millipore Supplier No. 32872100MG

Alanycarb is classified under the oxime carbamate group of insecticides. Its mode of action involves the inhibition of cholinesterase enzyme.

Agriculture

Specifications

Product Line PESTANAL
CAS 83130-01-2
Physical Form Neat
Linear Formula C17H25N3O4S2
MDL Number MFCD00274585
Type Analytical Standard
Quantity 100 mg
Formula Weight 399.53
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH