missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Alachlor ESA sodium salt, MilliporeSigma™ Supelco™

Catalog No. 111013063
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013063 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013063 Supplier Merck Emd Millipore Supplier No. 3414710MG

Alachlor ESA sodium salt is a metabolite of the preemergence herbicide, alachlor used to control annual grasses and broad-leaved weeds in corn and other crops.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 140939-15-7
Physical Form Neat
Linear Formula C14H20 NNaO5 S
MDL Number MFCD09037344
Synonym 2-[(2,6-Diethylphenyl)(methoxymethyl)-amino]-2-oxo-ethanesulfonic acid sodium salt
Type Analytical Standard
Quantity 10 mg
Formula Weight 337.37
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH