missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Acifluorfen, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 50594-66-6 |
| Physical Form | Neat |
| Linear Formula | C14H7 ClF3 NO5 |
| MDL Number | MFCD00143541 |
| Synonym | 5-(2-Chloro-alpha,alpha,alpha-trifluoro-p-tolyloxy)-2-nitrobenzoic acid; 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 361.66 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH