missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Acequinocyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 57960-19-7 |
| Physical Form | Neat |
| Linear Formula | C24H32O4 |
| MDL Number | MFCD01938252 |
| Synonym | 2-(Acetyloxy)-3-dodecyl-1,4-naphthalenedione; 3-Acetoxy-2-dodecyl-1,4-naphthoquinone; 3-Dodecyl-1,4-dihydro-1,4-dioxo-2-naphthyl acetate |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 384.51 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH