missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 4-tert-Butylphenol, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111020644
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020644 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020644 Supplier Merck Emd Millipore Supplier No. 506761

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 98-54-4
Product Line PESTANAL
Vapor Pressure 1 mmHg (70°C)
Density 0.908 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)3 CC6H4OH
MDL Number MFCD00002367
Melting Point 96°C to 101°C
Boiling Point 236°C to 238°C
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 150.22
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH