missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 4-Isopropylthioxanthone, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 83846-86-0 |
| Physical Form | Neat |
| Linear Formula | C16H14 OS |
| MDL Number | MFCD00192247 |
| Synonym | 4-Isopropyl-thioxanthene-9-one |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 254.35 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH