missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 4-Chlorophenoxyacetic acid, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 122-88-3 |
| Physical Form | Neat |
| Linear Formula | C8H7 ClO3 |
| MDL Number | MFCD00004305 |
| Synonym | 4-CPA |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 186.59 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH