missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 4,4-DDMU, MilliporeSigma™ Supelco™

Catalog No. 111019045
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019045 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019045 Supplier Merck Emd Millipore Supplier No. 35489250MG
Agriculture

Specifications

Product Line PESTANAL
CAS 1022-22-6
Flash Point >100°C
Physical Form Neat
Linear Formula C14H9 Cl3
MDL Number MFCD00055275
Synonym 2-Chloro-1,1-bis(4-chlorophenyl)-ethene
Type Analytical Standard
Quantity 250 mg
Formula Weight 283.58
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH