missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 4,4-DDE, MilliporeSigma™ Supelco™

Catalog No. 111019755
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019755 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019755 Supplier Merck Emd Millipore Supplier No. 35487100MG

4,4′-DDE can be found to be a breakdown product of trichloro-2,2-bis(4-chlorophenyl)ethane, in an aerobic environment. It can behave like a typical potent androgen receptor antagonist.

Agriculture

Specifications

Product Line PESTANAL
CAS 72-55-9
Physical Form Neat
Linear Formula (ClC6H4)2 C=CCl2
MDL Number MFCD00000837
Melting Point 88°C to 90°C
Synonym 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene; 4,4 ′-DDE
Type Analytical Standard
Quantity 100 mg
Formula Weight 318.03
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH