missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 3,5,6-Trichloro-2-pyridinol, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 6515-38-4 |
| Physical Form | Neat |
| Linear Formula | C5H2 Cl3 NO |
| MDL Number | MFCD00130269 |
| Synonym | 2,3,5-Trichloro-6-hydroxypyridine; 3,5,6-Trichloro-2-pyridone; TCP |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 198.43 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH