missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2-(Octylsulfanyl)ethan-1-ol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018328
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018328 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018328 Supplier Merck Emd Millipore Supplier No. 9252925MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 3547-33-9
Product Line PESTANAL
Linear Formula C10H22 OS
MDL Number MFCD00014046
Synonym 2-(Octylthio)ethanol; 2-Hydroxyethyl octyl sulfide
Type Analytical Standard
Quantity 25 mg
Formula Weight 190.35
Percent Purity ≥98% (GC)
Grade PESTANAL™ Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH