missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2-Imidazolidinethione, MilliporeSigma™ Supelco™

Catalog No. 111020295
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020295 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020295 Supplier Merck Emd Millipore Supplier No. 45531250MG
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 96-45-7
Physical Form Neat
Linear Formula C3H6N2 S
MDL Number MFCD00005276
Melting Point 196°C to 200°C
Synonym 2-Thioxoimidazolidine; N,N′-Ethylenethiourea
Type Analytical Standard
Quantity 250 mg
Formula Weight 102.16
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH