missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2,6-Dimethylphenol, MilliporeSigma™ Supelco™

Catalog No. 111020486
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020486 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020486 Supplier Merck Emd Millipore Supplier No. 367151G
Only null left

2,6-Dimethylphenol belongs to the class of phenols, commonly present in cigarette smoke and is a major toxic compound related to environmental and health issues.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
Ignition Point 1110°F
CAS 576-26-1
Flash Point 86°C - Closed Cup
Physical Form Neat
Linear Formula (CH3)2C6H3 OH
MDL Number MFCD00002240
Melting Point 43°C to 45°C
Boiling Point 203°C
Synonym 2-Hydroxy-m-xylene; vic.-m-Xylenol
Type Analytical Standard
Quantity 1 g
Formula Weight 122.16
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH