missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2,4-D, MilliporeSigma™ Supelco™

Catalog No. 111019789
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019789 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019789 Supplier Merck Emd Millipore Supplier No. 49083

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,4-D is a widely used, colorless and odorless herbicide, which can find applications in controlling broad-leaved weeds, woody plants in agriculture and also broad-leaves aquatic weeds. It can readily undergo degradation by microorganisms in soil and water.

Agriculture and Environmental

Specifications

Packaging Vial
Product Line PESTANAL
CAS 94-75-7
Vapor Pressure 0.4 mmHg (160°C)
Physical Form Neat
Linear Formula Cl2C6H3 OCH2 CO2 H
MDL Number MFCD00004300
Melting Point 136°C to 140°C
Synonym 2,4-Dichlorophenoxyacetic acid; 2,4-D
Type Analytical Standard
Quantity 1000 mg
Formula Weight 221.04
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH