missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2,4,6-Tribromanisole Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022457
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022457 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022457 Supplier Merck Emd Millipore Supplier No. 33489100MGR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 607-99-8
Melting Point 84°C to 88°C (literature)
Boiling Point 297°C to 299°C (literature)
Molecular Formula C7H5Br3O
Linear Formula Br3C6H2OCH3
Quantity 100 mg
MDL Number MFCD00192510
UN Number NONH for all modes of transport
Synonym 2,4,6-Tribromo-1-methoxybenzene
Formula Weight 344.83
Grade Analytical Standard
Physical Form Neat
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH