missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 2,3,4-Trichloroanisole, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 54135-80-7 |
| Physical Form | Neat |
| Linear Formula | C7H5 Cl3 O |
| MDL Number | MFCD00000541 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 211.47 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH