missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 1,2-Dibromo-3-chloropropane, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 96-12-8 |
| Density | 2.081 g/mL (at 25°C) |
| Flash Point | 76.7°C |
| Physical Form | Neat |
| Linear Formula | CH2 BrCHBrCH2 Cl |
| MDL Number | MFCD00039365 |
| Boiling Point | 60°C to 65°C (7.5 mmHg) |
| Synonym | DBCP |
| Type | Analytical Standard |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH