missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Perylene | |
| 279°C | |
| MFCD00004142 | |
| peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl | |
| C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2 | |
| 252.316 | |
| CHEBI:29861 | |
| ≥98.0% (GC) |
| 198-55-0 | |
| C20H12 | |
| 25 g | |
| CSHWQDPOILHKBI-UHFFFAOYSA-N | |
| perylene | |
| 9142 | |
| 252.32 | |
| Crystalline Powder |
Chemical Identifiers
| 198-55-0 | |
| 252.316 | |
| CSHWQDPOILHKBI-UHFFFAOYSA-N | |
| 9142 | |
| perylene |
| C20H12 | |
| MFCD00004142 | |
| peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl | |
| CHEBI:29861 | |
| C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2 |
Safety and Handling
RTECSNumber : SE3794000
TSCA : Yes